Assay Details
| Gene Symbol: Dus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301176.1 | NP_001288105.1 | 5 - 6 | 441 | 97 | |
| NM_025518.4 | NP_079794.1 | 6 - 7 | 485 | 97 | ||
| GenBank mRNA | AK009391.1 | - | 6 - 7 | 457 | 97 | |
| AK077451.1 | - | 5 - 6 | 413 | 97 | ||
| BC116934.1 | - | 6 - 7 | 403 | 40126253 | 97 | |
| BC116936.1 | - | 6 - 7 | 403 | 40126255 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001301176.1 transcripts.
Set Membership: |
|

Dus2
Genomic Map