Assay Details
| Gene Symbol: Dus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301176.1 | NP_001288105.1 | 6 - 7 | 488 | 72 | |
| NM_025518.4 | NP_079794.1 | 7 - 8 | 532 | 72 | ||
| GenBank mRNA | AK009391.1 | - | 7 - 8 | 504 | 72 | |
| AK077451.1 | - | 6 - 7 | 460 | 72 | ||
| BC116934.1 | - | 7 - 8 | 450 | 40126253 | 72 | |
| BC116936.1 | - | 7 - 8 | 450 | 40126255 | 72 | |
More Information
Set Membership: |
|

Dus2
Genomic Map