Assay Details
Gene Symbol: Sgk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291152.1 | NP_001278081.1 | 5 - 6 | 501 | 83 | |
GenBank mRNA | BC026549.1 | - | 4 - 5 | 246 | 4973908 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
