Assay Details
| Gene Symbol: Ano4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277188.1 | NP_001264117.1 | 17 - 18 | 2194 | 88 | |
| NR_102339.1 | 16 - 17 | 2151 | 88 | |||
| GenBank mRNA | BC065114.1 | - | 16 - 17 | 2151 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ano4
Genomic Map