Assay Details
| Gene Symbol: Dus1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026824.4 | NP_081100.2 | 4 - 5 | 660 | 62 | |
| GenBank mRNA | AK004041.1 | - | 3 - 4 | 183 | 62 | |
| AK037538.1 | - | 3 - 4 | 1074 | 62 | ||
| AK088243.1 | - | 4 - 5 | 661 | 62 | ||
| AK147967.1 | - | 4 - 5 | 859 | 62 | ||
| AK148307.1 | - | 4 - 5 | 859 | 62 | ||
| BC019480.1 | - | 2 - 3 | 144 | 4037583 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dus1l
Genomic Map