Assay Details
| Gene Symbol: Dus1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026824.4 | NP_081100.2 | 10 - 11 | 1292 | 63 | |
| GenBank mRNA | AK004041.1 | - | 9 - 10 | 815 | 63 | |
| AK037538.1 | - | 8 - 9 | 2429 | 63 | ||
| AK049333.1 | - | 163 | 63 | |||
| AK088243.1 | - | 10 - 11 | 1293 | 63 | ||
| AK147967.1 | - | 10 - 11 | 1492 | 63 | ||
| AK148307.1 | - | 10 - 11 | 1492 | 63 | ||
| BC019480.1 | - | 8 - 9 | 776 | 4037583 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dus1l
Genomic Map