Assay Details
| Gene Symbol: Tmem104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033393.2 | NP_001028565.1 | 5 - 6 | 527 | 72 | |
| GenBank mRNA | AK040894.1 | - | 5 - 6 | 489 | 72 | |
| AK049868.1 | - | 5 - 6 | 464 | 72 | ||
| AK131112.1 | - | 5 - 6 | 504 | 72 | ||
| AK155041.1 | - | 5 - 6 | 504 | 72 | ||
| AK167098.1 | - | 5 - 6 | 483 | 72 | ||
| AK171314.1 | - | 5 - 6 | 504 | 72 | ||
| AK171457.1 | - | 5 - 6 | 489 | 72 | ||
| BC140976.1 | - | 5 - 6 | 447 | 9055588 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem104
Genomic Map