Assay Details
| Gene Symbol: Tmem104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033393.2 | NP_001028565.1 | 7 - 8 | 709 | 55 | |
| GenBank mRNA | AK049868.1 | - | 7 - 8 | 646 | 55 | |
| AK131112.1 | - | 7 - 8 | 686 | 55 | ||
| AK155041.1 | - | 7 - 8 | 686 | 55 | ||
| AK167098.1 | - | 7 - 8 | 665 | 55 | ||
| AK171314.1 | - | 7 - 8 | 686 | 55 | ||
| AK171457.1 | - | 7 - 8 | 671 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem104
Genomic Map