Assay Details
| Gene Symbol: Usp36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033528.1 | NP_001028700.1 | 3 - 4 | 649 | 66 | |
| GenBank mRNA | AK028572.1 | - | 3 - 4 | 694 | 66 | |
| AK049205.1 | - | 3 - 4 | 479 | 66 | ||
| AK079342.1 | - | 3 - 4 | 534 | 66 | ||
| AK129363.1 | - | 3 - 4 | 447 | 66 | ||
| AK165635.1 | - | 3 - 4 | 597 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
ubiquitin-dependent protein catabolic process
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
regulation of mitophagy
positive regulation of protein targeting to mitochondrion
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
peptidase activity
cysteine-type peptidase activity
hydrolase activity
thiol-dependent ubiquitinyl hydrolase activity
poly(A) RNA binding
proteolysis
ubiquitin-dependent protein catabolic process
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
regulation of mitophagy
positive regulation of protein targeting to mitochondrion
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
peptidase activity
cysteine-type peptidase activity
hydrolase activity
thiol-dependent ubiquitinyl hydrolase activity
poly(A) RNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2133767 | AK129363.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2133767 | AK165635.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2133767 | AK079342.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2133767 | AK028572.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2133767 | NM_001033528.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P627925 | AK129363.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P627925 | AK049205.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P627925 | AK028915.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P627925 | AK028572.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P627925 | NM_001033528.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P627925 | AK129363.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P627925 | AK049205.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P627925 | AK028915.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P627925 | AK028572.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P627925 | NM_001033528.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P02321 | AK028915.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P02321 | NM_001033528.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 449862 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323121 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 501412 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215187 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 38742 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737850 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 928458 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674021 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 119766 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489032 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537031 | AK002372.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537031 | AK003073.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537031 | AK004150.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537031 | BC058123.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537031 | XM_901690.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537031 | XM_903949.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537031 | XM_922038.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606230 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139246 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 772599 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19637 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30016 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 120592 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96355 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28207 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164233 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161562 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165550 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2133767 | AK129363.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2133767 | AK165635.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2133767 | AK079342.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2133767 | AK028572.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2133767 | NM_001033528.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P627925 | AK129363.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P627925 | AK049205.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P627925 | AK028915.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P627925 | AK028572.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P627925 | NM_001033528.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P627925 | AK129363.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P627925 | AK049205.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P627925 | AK028915.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P627925 | AK028572.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P627925 | NM_001033528.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P02321 | AK028915.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P02321 | NM_001033528.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161562 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165550 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164233 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02442239 | - | - |
| Whole Mouse Genome G4122A | A_51_P119016 | AK049205.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P119016 | AK047554.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P119016 | AK028572.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P119016 | NM_001033528.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627925 | AK129363.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627925 | AK049205.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627925 | AK028915.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627925 | AK028572.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627925 | NM_001033528.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627925 | AK129363.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627925 | AK049205.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627925 | AK028915.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627925 | AK028572.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P627925 | NM_001033528.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161562 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164233 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09276669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09092202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09725124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09405504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09725141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08998533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09092209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09055458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09092207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09405505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09276684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08998553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08998551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08998535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09276665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08998541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09405502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09725131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09697251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09594915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09594917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28170387 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10393527 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393518 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393516 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393519 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393525 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393524 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393513 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393517 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393514 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393520 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393529 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393528 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393531 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393511 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393512 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393510 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393515 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393530 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393521 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393522 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393526 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456094_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458311_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455237_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10393509 | - | - |
| Mouse Genome 430 2.0 Array | 1455237_at | - | - |
| Mouse Genome 430 2.0 Array | 1456094_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455237_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458311_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456094_at | - | - |
| Mouse Genome 430 2.0 Array | 1458311_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72344_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72344_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100039508.mm.1 | NM_001033528.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039508.mm.1 | NM_001033528.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039508.mm.1 | TC1100004156.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100039512.mm.1 | NM_001033528.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039512.mm.1 | NM_001033528.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039512.mm.1 | TC1100004156.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100039514.mm.1 | NM_001033528.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039514.mm.1 | NM_001033528.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039514.mm.1 | TC1100004156.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100039510.mm.1 | NM_001033528.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039510.mm.1 | NM_001033528.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039510.mm.1 | TC1100004156.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100039507.mm.1 | NM_001033528.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039507.mm.1 | NM_001033528.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039507.mm.1 | TC1100004156.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100039513.mm.1 | NM_001033528.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039513.mm.1 | NM_001033528.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039513.mm.1 | TC1100004156.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100039509.mm.1 | NM_001033528.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039509.mm.1 | NM_001033528.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039509.mm.1 | TC1100004156.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455237_PM_at | - | - |
| Mouse Expression Set 430 | 1456094_at | - | - |
| Mouse Expression Set 430 | 1455237_at | - | - |
| Mouse Expression Set 430 | 1456094_PM_at | - | - |
| Mouse Expression Set 430 | 1458311_at | - | - |
| Mouse Expression Set 430 | 1458311_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455237_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456094_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455237_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456094_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456094_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455237_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455237_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456094_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458311_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458311_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 557925 | - | - |
| ABI Mouse Genome Survey Microarray | 786723 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 100110408 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2491232 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224255 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2491232 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244609 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100110408 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100110408 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244609 | - | - |

Usp36
Genomic Map