Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800007553.mm.1 |
NM_001001493.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007553.mm.1 |
NM_001001493.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007553.mm.1 |
TC0800000982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800007552.mm.1 |
NM_001001493.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007552.mm.1 |
NM_001001493.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007552.mm.1 |
TC0800000982.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800007550.mm.1 |
NM_001001493.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007550.mm.1 |
NM_001001493.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800007550.mm.1 |
TC0800000982.mm.1 |
MTA TC |