Assay Details
| Gene Symbol: Wdr83os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001493.2 | NP_001001493.1 | 3 - 4 | 311 | 76 | |
| GenBank mRNA | AK010893.1 | - | 3 - 4 | 313 | 76 | |
| AK147129.1 | - | 3 - 4 | 535 | 76 | ||
| BC024937.1 | - | 3 - 4 | 308 | 76 | ||
| BC056474.1 | - | 3 - 4 | 281 | 6823629 | 76 | |
| BC089623.1 | - | 3 - 4 | 311 | 30118707 | 76 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046907 | BC056474.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046907 | BC089623.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046907 | BC091742.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046907 | NM_001001493.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046907 | AK147129.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960506 | BC089623.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960506 | NM_001001493.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960506 | AK147129.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157812 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960506 | BC089623.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960506 | NM_001001493.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960506 | AK147129.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046907 | BC056474.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046907 | BC089623.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046907 | BC091742.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046907 | NM_001001493.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046907 | AK147129.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26023623 | - | - |
| Whole Mouse Genome G4122A | A_51_P307062 | BC056474.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307062 | BC089623.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307062 | BC091742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307062 | NM_001001493.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307062 | AK147129.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P245853 | BC089623.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P245853 | NM_001001493.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P245853 | AK010893.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P245853 | AK147129.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P245853 | BC024937.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106648 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140211 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07099620 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07099626 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07099607 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07099612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26023663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07099626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07099607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26023619 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10573582 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573580 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573581 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424117_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10573578 | - | - |
| Mouse Genome 430 2.0 Array | 1424117_at | - | - |
| Mouse Genome 430 2.0 Array | 1424117_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 414077_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 414077_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800007555.mm.1 | NM_001001493.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007555.mm.1 | NM_001001493.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007555.mm.1 | TC0800000982.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800007554.mm.1 | NM_001001493.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007554.mm.1 | NM_001001493.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800007554.mm.1 | TC0800000982.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424117_PM_at | - | - |
| Mouse Expression Set 430 | 1424117_at | - | - |
| Murine Genome U74v2 Set | 98516_PM_at | - | - |
| Murine Genome U74v2 Set | 98516_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424117_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424117_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424117_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424117_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 331532 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101090181 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101090181 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2546540 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101090181 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101090181 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2939185 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2939185 | - | - |

Wdr83os
Genomic Map