Assay Details
Gene Symbol: C230057M02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK032654.1 | - | 2 - 3 | 959 | 59 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
core-binding factor, runt domain, alpha subunit 2, translocated to, 3 (human) |
Cbfa2t3 |
A630044F12Rik, AI465270, AW229127, Cbfa2t3h, ETO-2, Eto2, MTGR2 |
-36.1kb | Sense | Chr. 8 - 122625136 - 122699639 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA A530010L16 gene | A530010L16Rik |
BC032935, Gm1908 |
23.6kb | Sense | Chr. 8 - 122760034 - 122762874 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
