Assay Details
| Gene Symbol: Cse1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023565.3 | NP_076054.1 | 18 - 19 | 2059 | 128 | |
| GenBank mRNA | AF301152.1 | - | 18 - 19 | 2059 | 128 | |
| AK138940.1 | - | 4 - 5 | 483 | 128 | ||
| BC145393.1 | - | 17 - 18 | 1830 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cse1l
Genomic Map