Assay Details
| Gene Symbol: Bmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007557.3 | NP_031583.2 | 1 - 2 | 661 | 65 | |
| GenBank mRNA | AK020411.1 | - | 1 - 2 | 433 | 65 | |
| BC010771.1 | - | 1 - 2 | 633 | 4218495 | 65 | |
| CJ094104.1 | - | 1 - 2 | 411 | 65 | ||
| X56906.1 | - | 1 - 2 | 512 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bmp7
Genomic Map