Assay Details
| Gene Symbol: Bmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007557.3 | NP_031583.2 | 4 - 5 | 1211 | 75 | |
| GenBank mRNA | BC010771.1 | - | 4 - 5 | 1183 | 4218495 | 75 |
| CA316109.1 | - | 2 - 3 | 345 | 75 | ||
| X56906.1 | - | 4 - 5 | 1062 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bmp7
Genomic Map