| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200019848.mm.1 |
NM_001159662.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019848.mm.1 |
NM_001159662.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019848.mm.1 |
NM_153089.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019848.mm.1 |
NM_153089.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019848.mm.1 |
TC0200002439.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200019847.mm.1 |
NM_001159662.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019847.mm.1 |
NM_001159662.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019847.mm.1 |
NM_153089.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019847.mm.1 |
NM_153089.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019847.mm.1 |
TC0200002439.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200019849.mm.1 |
NM_001159662.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019849.mm.1 |
NM_001159662.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019849.mm.1 |
NM_153089.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019849.mm.1 |
NM_153089.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019849.mm.1 |
TC0200002439.mm.1 |
MTA TC |