Assay Details
| Gene Symbol: Mmp24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010808.3 | NP_034938.3 | 4 - 5 | 868 | 57 | |
| GenBank mRNA | AB021226.1 | - | 4 - 5 | 813 | 57 | |
| AJ010262.1 | - | 4 - 5 | 814 | 57 | ||
| CD352405.1 | - | 1 - 2 | 65 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp24
Genomic Map