Assay Details
| Gene Symbol: Ptpn23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081043.1 | NP_001074512.1 | 6 - 7 | 638 | 87 | |
| GenBank mRNA | CF734421.1 | - | 1 - 2 | 137 | 87 | |
More Information
Set Membership: |
|

Ptpn23
Genomic Map