Assay Details
| Gene Symbol: Fam20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153782.1 | NP_722477.1 | 6 - 7 | 1465 | 70 | |
| GenBank mRNA | AK142932.1 | - | 6 - 7 | 1490 | 70 | |
| BC011274.1 | - | 6 - 7 | 1369 | 70 | ||
| BC029169.1 | - | 6 - 7 | 1465 | 4947019 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam20a
Genomic Map