Assay Details
| Gene Symbol: Slc7a6os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007567.2 | NP_001007568.1 | 4 - 5 | 812 | 67 | |
| GenBank mRNA | AK010254.1 | - | 4 - 5 | 807 | 67 | |
| AK146733.1 | - | 4 - 5 | 813 | 67 | ||
| AK171806.1 | - | 2 - 3 | 2526 | 67 | ||
| BC054118.1 | - | 4 - 5 | 810 | 30021749 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26138372 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26138373 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26138371 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26138375 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26138370 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26138369 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P371675 | BC054118.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P371675 | AK171806.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P371675 | AK010254.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P371675 | AK146733.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P371675 | NM_001007567.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P371675 | BC054118.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P371675 | AK171806.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P371675 | AK010254.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P371675 | AK146733.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P371675 | NM_001007567.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365216 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714699 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 100191 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 696160 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506828 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155635 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137592 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149572 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P371675 | BC054118.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P371675 | AK171806.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P371675 | AK010254.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P371675 | AK146733.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P371675 | NM_001007567.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P371675 | BC054118.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P371675 | AK171806.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P371675 | AK010254.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P371675 | AK146733.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P371675 | NM_001007567.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26138353 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137592 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06628910 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149572 | - | - |
| Whole Mouse Genome G4122A | A_51_P371675 | BC054118.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P371675 | AK171806.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P371675 | AK010254.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P371675 | AK146733.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P371675 | NM_001007567.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P371675 | BC054118.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P371675 | AK171806.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P371675 | AK010254.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P371675 | AK146733.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P371675 | NM_001007567.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137592 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149572 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06826995 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06826996 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26138372 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26138373 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26138371 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26138369 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26138370 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26138375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26138336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06716689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06745711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06745713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06833073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06786868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06870079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26138327 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581477 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581475 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581478 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581474 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581476 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429596_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440303_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581473 | - | - |
| Mouse Genome 430 2.0 Array | 1440303_at | - | - |
| Mouse Genome 430 2.0 Array | 1429596_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440303_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429596_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66432_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66432_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800022031.mm.1 | NM_001007567.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022031.mm.1 | NM_001007567.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022031.mm.1 | TC0800002854.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800022032.mm.1 | NM_001007567.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022032.mm.1 | NM_001007567.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022032.mm.1 | TC0800002854.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1440303_at | - | - |
| Mouse Expression Set 430 | 1429596_PM_at | - | - |
| Mouse Expression Set 430 | 1440303_PM_at | - | - |
| Mouse Expression Set 430 | 1429596_at | - | - |
| Murine Genome U74v2 Set | 165738_PM_r_at | - | - |
| Murine Genome U74v2 Set | 165738_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440303_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429596_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440303_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429596_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 493721 | - | - |
| ABI Mouse Genome Survey Microarray | 597902 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3162446 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3162446 | - | - |
| mouse-6 v1.1 (Illumina) | GI_57222271-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3162446 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3162446 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_57222271-S | - | - |

Slc7a6os
Genomic Map