Assay Details
| Gene Symbol: Slc41a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177388.3 | NP_796362.1 | 5 - 6 | 1118 | 57 | |
| GenBank mRNA | AK038549.1 | - | 5 - 6 | 1119 | 57 | |
| BC138146.1 | - | 4 - 5 | 956 | 8861165 | 57 | |
| BC138147.1 | - | 4 - 5 | 956 | 8861166 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc41a2
Genomic Map