Assay Details
| Gene Symbol: Cmtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181990.2 | NP_871790.2 | 3 - 4 | 924 | 145 | |
| GenBank mRNA | AY369078.2 | - | 471 | 145 | ||
| BC145587.1 | - | 3 - 4 | 924 | 9054153 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2078935 | AY369078.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2078935 | AY241874.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972464 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158594 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117489 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106376 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972464 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2078935 | AY369078.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2078935 | AY241874.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139488 | AY369078.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139488 | AY241873.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139488 | AY241874.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139488 | AY162281.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P139488 | AY172740.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127877 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06623710 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06623736 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06623672 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117489 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01816083 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158594 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06623661 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06623762 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06623758 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01816102 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106376 | - | - |
| Whole Mouse Genome G4122A | A_52_P139488 | AY369078.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P139488 | AY241873.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P139488 | AY241874.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P139488 | AY162281.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P139488 | AY172740.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06623672 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106376 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06623762 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06623710 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158594 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117489 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06756334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06883320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06937440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127843 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581059 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581058 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581052 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581054 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431710_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581049 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800021394.mm.1 | NM_181990.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021394.mm.1 | NM_181990.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021394.mm.1 | TC0800002798.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800021393.mm.1 | NM_181990.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021393.mm.1 | NM_181990.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021393.mm.1 | TC0800002798.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431710_PM_at | - | - |
| Mouse Expression Set 430 | 1431710_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431710_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431710_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 719400 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2260494 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2260494 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002260494 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2706847 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2706847 | - | - |
| mouse-6 v1.1 (Illumina) | scl0353509.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2260494 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0353509.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260494 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2809355 | - | - |

Cmtm1
Genomic Map