Assay Details
| Gene Symbol: Nudt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027722.4 | NP_081998.3 | 1 - 2 | 506 | 92 | |
| GenBank mRNA | AK017075.1 | - | 1 - 2 | 403 | 92 | |
| AK048062.1 | - | 1 - 2 | 486 | 92 | ||
| AK211623.1 | - | 1 - 2 | 106 | 92 | ||
| BC027209.1 | - | 1 - 2 | 167 | 3994391 | 92 | |
| CJ169842.1 | - | 2 - 3 | 301 | 92 | ||
| CT010342.1 | - | 1 - 2 | 96 | 92 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BC030004.1 | - | Chr. - - [-] on Build | 92 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Nudt4
Genomic Map