| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200019188.mm.1 |
NM_172674.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019188.mm.1 |
NM_172674.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019188.mm.1 |
TC0200002386.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200019189.mm.1 |
NM_172674.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019189.mm.1 |
NM_172674.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019189.mm.1 |
TC0200002386.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200019186.mm.1 |
NM_172674.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019186.mm.1 |
NM_172674.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019186.mm.1 |
TC0200002386.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200019191.mm.1 |
NM_172674.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019191.mm.1 |
NM_172674.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200019191.mm.1 |
TC0200002386.mm.1 |
MTA TC |