Assay Details
| Gene Symbol: Farsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025648.3 | NP_079924.2 | 10 - 11 | 1248 | 57 | |
| GenBank mRNA | AK002577.1 | - | 10 - 11 | 1223 | 57 | |
| AK004463.1 | - | 1 - 2 | 136 | 57 | ||
| AK016506.1 | - | 9 - 10 | 1345 | 57 | ||
| AK031697.1 | - | 10 - 11 | 1226 | 57 | ||
| AK154629.1 | - | 10 - 11 | 1246 | 57 | ||
| AK171872.1 | - | 10 - 11 | 1128 | 57 | ||
| AK204847.1 | - | 1 - 2 | 97 | 57 | ||
| AK210915.1 | - | 1 - 2 | 97 | 57 | ||
| AK214616.1 | - | 1 - 2 | 97 | 57 | ||
| BC006862.1 | - | 10 - 11 | 1219 | 3595108 | 57 | |
| BC013533.1 | - | 10 - 11 | 1214 | 4238911 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Farsa
Genomic Map