Assay Details
| Gene Symbol: Farsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025648.3 | NP_079924.2 | 6 - 7 | 771 | 83 | |
| GenBank mRNA | AK002577.1 | - | 6 - 7 | 748 | 83 | |
| AK016506.1 | - | 6 - 7 | 767 | 83 | ||
| AK031697.1 | - | 6 - 7 | 749 | 83 | ||
| AK044637.1 | - | 6 - 7 | 749 | 83 | ||
| AK084031.1 | - | 6 - 7 | 733 | 83 | ||
| AK154629.1 | - | 6 - 7 | 769 | 83 | ||
| AK171872.1 | - | 6 - 7 | 651 | 83 | ||
| BC006862.1 | - | 6 - 7 | 742 | 3595108 | 83 | |
| BC013533.1 | - | 6 - 7 | 740 | 4238911 | 83 | |
| CT010350.1 | - | 6 - 7 | 722 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Farsa
Genomic Map