Assay Details
| Gene Symbol: Farsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025648.3 | NP_079924.2 | 7 - 8 | 890 | 85 | |
| GenBank mRNA | AK002577.1 | - | 7 - 8 | 867 | 84 | |
| AK016506.1 | - | 7 - 8 | 886 | 85 | ||
| AK031697.1 | - | 7 - 8 | 868 | 85 | ||
| AK154629.1 | - | 7 - 8 | 888 | 85 | ||
| AK171872.1 | - | 7 - 8 | 770 | 85 | ||
| BC006862.1 | - | 7 - 8 | 861 | 3595108 | 85 | |
| BC013533.1 | - | 7 - 8 | 859 | 4238911 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Farsa
Genomic Map