Assay Details
| Gene Symbol: Farsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025648.3 | NP_079924.2 | 9 - 10 | 1071 | 83 | |
| GenBank mRNA | AK002577.1 | - | 9 - 10 | 1046 | 83 | |
| AK031697.1 | - | 9 - 10 | 1049 | 83 | ||
| AK154629.1 | - | 9 - 10 | 1069 | 83 | ||
| AK171872.1 | - | 9 - 10 | 951 | 83 | ||
| BC006862.1 | - | 9 - 10 | 1042 | 3595108 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Farsa
Genomic Map