Assay Details
| Gene Symbol: Gga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252067.1 | NP_001238996.1 | 7 - 8 | 659 | 93 | |
| NM_173048.3 | NP_766636.2 | 7 - 8 | 659 | 93 | ||
| GenBank mRNA | AK031086.1 | - | 7 - 8 | 631 | 93 | |
| AK082327.1 | - | 7 - 8 | 623 | 93 | ||
| AK122212.2 | - | 7 - 8 | 628 | 93 | ||
| AK132183.1 | - | 7 - 8 | 627 | 93 | ||
| AK147882.1 | - | 7 - 8 | 628 | 93 | ||
| AK216206.1 | - | 3 - 4 | 211 | 93 | ||
| BC024266.1 | - | 1266 | 93 | |||
| BC056424.1 | - | 7 - 8 | 613 | 93 | ||
| BC085617.1 | - | 7 - 8 | 638 | 93 | ||
| BC110676.1 | - | 7 - 8 | 615 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gga3
Genomic Map