Assay Details
| Gene Symbol: Tmem104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033393.2 | NP_001028565.1 | 1 - 2 | 107 | 76 | |
| GenBank mRNA | AK040894.1 | - | 1 - 2 | 69 | 76 | |
| AK049868.1 | - | 1 - 2 | 44 | 76 | ||
| AK131112.1 | - | 1 - 2 | 84 | 76 | ||
| AK155041.1 | - | 1 - 2 | 84 | 76 | ||
| AK167098.1 | - | 1 - 2 | 63 | 76 | ||
| AK171314.1 | - | 1 - 2 | 84 | 76 | ||
| AK171457.1 | - | 1 - 2 | 69 | 76 | ||
| BC140976.1 | - | 1 - 2 | 27 | 9055588 | 76 | |
| BY091433.1 | - | 1 - 2 | 108 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK155041.1, AK171457.1, BC140976.1, NM_001033393.2 transcripts.
Set Membership: |
|

Tmem104
Genomic Map