Assay Details
| Gene Symbol: Slc16a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030696.3 | NP_109621.1 | 1 - 2 | 95 | 80 | |
| GenBank mRNA | BC046525.1 | - | 1 - 2 | 95 | 5319754 | 80 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC046525.1, NM_030696.3 transcripts.
Set Membership: |
|

Slc16a3
Genomic Map