| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100029464.mm.1 |
NM_013625.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029464.mm.1 |
NM_013625.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029464.mm.1 |
NR_037610.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029464.mm.1 |
NR_037610.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029464.mm.1 |
TC1100003198.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100029465.mm.1 |
NM_013625.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029465.mm.1 |
NM_013625.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029465.mm.1 |
NR_037610.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029465.mm.1 |
NR_037610.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029465.mm.1 |
TC1100003198.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100029466.mm.1 |
NM_013625.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029466.mm.1 |
NM_013625.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029466.mm.1 |
NR_037610.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029466.mm.1 |
NR_037610.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029466.mm.1 |
TC1100003198.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100029461.mm.1 |
NM_013625.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029461.mm.1 |
NM_013625.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029461.mm.1 |
NR_037610.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029461.mm.1 |
NR_037610.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029461.mm.1 |
TC1100003198.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100029462.mm.1 |
NM_013625.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029462.mm.1 |
NM_013625.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029462.mm.1 |
NR_037610.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029462.mm.1 |
NR_037610.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029462.mm.1 |
TC1100003198.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100029471.mm.1 |
NR_037610.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029471.mm.1 |
NR_037610.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029471.mm.1 |
TC1100003198.mm.1 |
MTA TC |