Assay Details
Gene Symbol: Psmd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010817.2 | NP_034947.1 | 1 - 2 | 201 | 62 | |
GenBank mRNA | AK031013.1 | - | 1 - 2 | 152 | 62 | |
AK146128.1 | - | 149 | 62 | |||
AK159825.1 | - | 1 - 2 | 153 | 62 | ||
BC013513.1 | - | 1 - 2 | 183 | 62 | ||
BC130014.1 | - | 1 - 2 | 178 | 40140405 | 62 | |
M64641.1 | - | 1 - 2 | 149 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
