Assay Details
| Gene Symbol: 1700019N19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026208.2 | NP_080484.1 | 4 - 5 | 494 | 65 | |
| GenBank mRNA | AK006135.1 | - | 4 - 5 | 495 | 65 | |
| AK006423.1 | - | 4 - 5 | 491 | 65 | ||
| BC049561.1 | - | 4 - 5 | 482 | 6773246 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P307864 | NM_026208.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P307864 | AK006135.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P307864 | NM_026208.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P307864 | AK006135.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907647 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59268 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659862 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 354206 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 531478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 97856 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107454 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P307864 | NM_026208.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P307864 | AK006135.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P307864 | NM_026208.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P307864 | AK006135.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32215371 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107454 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09215762 | - | - |
| Whole Mouse Genome G4122A | A_51_P307858 | NM_026208.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307858 | AK006135.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307858 | AK006423.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307858 | BC049561.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307864 | NM_026208.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307864 | AK006135.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307864 | NM_026208.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307864 | AK006135.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109254 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139154 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107454 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14372845 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14372857 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14243676 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14243648 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14243671 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14243659 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14372856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32215281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14157560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14210515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14243668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215348 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14482973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215346 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14452098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215342 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14372850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14372848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14243676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14315203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14243671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32215278 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10468744 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468745 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468739 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468742 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468743 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468741 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468740 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451001_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10468738 | - | - |
| Mouse Genome 430 2.0 Array | 1451001_at | - | - |
| Mouse Genome 430 2.0 Array | 1451001_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67507_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67507_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900015274.mm.1 | NM_026208.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015274.mm.1 | NM_026208.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015274.mm.1 | TC1900001724.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900015272.mm.1 | NM_026208.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015272.mm.1 | NM_026208.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015272.mm.1 | TC1900001724.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900015273.mm.1 | NM_026208.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015273.mm.1 | NM_026208.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015273.mm.1 | TC1900001724.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900015275.mm.1 | NM_026208.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015275.mm.1 | NM_026208.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015275.mm.1 | TC1900001724.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451001_at | - | - |
| Mouse Expression Set 430 | 1451001_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451001_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451001_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451001_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451001_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 336459 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4050019 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004050019 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2618910 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2618910 | - | - |
| mouse-6 v1.1 (Illumina) | scl52333.7.1_104-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4050019 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52333.7.1_104-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4050019 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2618910 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2618910 | - | - |

1700019N19Rik
Genomic Map