Assay Details
| Gene Symbol: Gpatch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026181.1 | NP_080457.1 | 17 - 18 | 2545 | 65 | |
| GenBank mRNA | AK004868.1 | - | 17 - 18 | 2545 | 65 | |
| AK014367.1 | - | 17 - 18 | 2524 | 65 | ||
| AK154571.1 | - | 17 - 18 | 2525 | 65 | ||
| BC026394.1 | - | 11 - 12 | 1711 | 65 | ||
| BC100577.1 | - | 17 - 18 | 2520 | 65 | ||
| BC125584.1 | - | 17 - 18 | 2536 | 40130099 | 65 | |
| BC132155.1 | - | 17 - 18 | 2536 | 40130432 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpatch1
Genomic Map