Assay Details
| Gene Symbol: Lsm14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025948.2 | NP_080224.1 | 3 - 4 | 479 | 66 | |
| GenBank mRNA | AK003611.1 | - | 1 - 2 | 39 | 66 | |
| AK012276.1 | - | 2 - 3 | 242 | 66 | ||
| AK189673.1 | - | 2 - 3 | 214 | 66 | ||
| BC031521.1 | - | 3 - 4 | 479 | 2655161 | 66 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lsm14a
Genomic Map