Assay Details
| Gene Symbol: Ica1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027407.3 | NP_081683.3 | 6 - 7 | 979 | 80 | |
| GenBank mRNA | AK006525.1 | - | 4 - 5 | 507 | 80 | |
| AK029991.1 | - | 6 - 7 | 751 | 80 | ||
| AK158859.1 | - | 6 - 7 | 815 | 80 | ||
| BC058983.1 | - | 1 - 2 | 108 | 80 | ||
| BC079669.1 | - | 6 - 7 | 979 | 30355976 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ica1l
Genomic Map