Assay Details
| Gene Symbol: Cdk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033373.2 | NP_001028545.2 | 6 - 7 | 661 | 58 | |
| GenBank mRNA | BC147314.1 | - | 6 - 7 | 661 | 9056364 | 58 |
| BC147315.1 | - | 6 - 7 | 661 | 9056370 | 58 | |
| BC147758.1 | - | 6 - 7 | 661 | 9087483 | 58 | |
| BC147763.1 | - | 6 - 7 | 661 | 9087488 | 58 | |
| BC172036.1 | - | 6 - 7 | 658 | 9054322 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdk15
Genomic Map