Assay Details
| Gene Symbol: Cdk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033373.2 | NP_001028545.2 | 9 - 10 | 1008 | 62 | |
| GenBank mRNA | AK042881.1 | - | 2 - 3 | 217 | 62 | |
| AK211120.1 | - | 2 - 3 | 62 | |||
| AK217269.1 | - | 3 - 4 | 334 | 62 | ||
| BC147314.1 | - | 9 - 10 | 1008 | 9056364 | 62 | |
| BC147315.1 | - | 9 - 10 | 1008 | 9056370 | 62 | |
| BC147758.1 | - | 9 - 10 | 1008 | 9087483 | 62 | |
| BC147763.1 | - | 9 - 10 | 1008 | 9087488 | 62 | |
| BC172036.1 | - | 9 - 10 | 1005 | 9054322 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdk15
Genomic Map