Assay Details
| Gene Symbol: Mettl21e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207281.3 | NP_997164.1 | 1 - 2 | 89 | 155 | |
| GenBank mRNA | AK029331.1 | - | 1 - 2 | 90 | 155 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029331.1, NM_207281.3 transcripts.
Set Membership: |
|

Mettl21e
Genomic Map