| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100018342.mm.1 |
NM_207281.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018342.mm.1 |
NM_207281.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018342.mm.1 |
TC0100002274.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100018340.mm.1 |
NM_207281.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018340.mm.1 |
NM_207281.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018340.mm.1 |
TC0100002274.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100018341.mm.1 |
NM_207281.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018341.mm.1 |
NM_207281.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018341.mm.1 |
TC0100002274.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100018343.mm.1 |
NM_207281.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018343.mm.1 |
NM_207281.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100018343.mm.1 |
TC0100002274.mm.1 |
MTA TC |