Assay Details
| Gene Symbol: Rpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025683.2 | NP_079959.2 | 1 - 2 | 136 | 66 | |
| GenBank mRNA | AK007735.1 | - | 1 - 2 | 142 | 66 | |
| AK017774.1 | - | 1 - 2 | 137 | 66 | ||
| AK088152.1 | - | 1 - 2 | 137 | 66 | ||
| AK150008.1 | - | 1 - 2 | 174 | 66 | ||
| AK185217.1 | - | 1 - 2 | 130 | 66 | ||
| BC006953.1 | - | 1 - 2 | 130 | 66 | ||
| BC019126.1 | - | 1 - 2 | 179 | 5008075 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rpe
Genomic Map