| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600013975.mm.1 |
NM_178797.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600013975.mm.1 |
NM_178797.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600013975.mm.1 |
TC0600001751.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600013972.mm.1 |
NM_178797.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600013972.mm.1 |
NM_178797.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600013972.mm.1 |
TC0600001751.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600013973.mm.1 |
NM_178797.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600013973.mm.1 |
NM_178797.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600013973.mm.1 |
TC0600001751.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600013974.mm.1 |
NM_178797.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600013974.mm.1 |
NM_178797.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600013974.mm.1 |
TC0600001751.mm.1 |
MTA TC |