Assay Details
| Gene Symbol: St8sia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013666.2 | NP_038694.2 | 3 - 4 | 574 | 71 | |
| NM_153124.2 | NP_694764.2 | 2 - 3 | 466 | 71 | ||
| GenBank mRNA | AK013542.1 | - | 2 - 3 | 469 | 71 | |
| AK078670.1 | - | 2 - 3 | 249 | 71 | ||
| AK162549.1 | - | 2 - 3 | 246 | 71 | ||
| BC034855.1 | - | 2 - 3 | 460 | 5146703 | 71 | |
| BY733136.1 | - | 3 - 4 | 556 | 71 | ||
| X98014.1 | - | 3 - 4 | 437 | 71 | ||
More Information
Set Membership: |
|

St8sia5
Genomic Map