| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400033329.mm.1 |
NM_001003898.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033329.mm.1 |
NM_001003898.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033329.mm.1 |
NM_001003899.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033329.mm.1 |
NM_001003899.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033329.mm.1 |
NM_001008545.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033329.mm.1 |
NM_001008545.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033329.mm.1 |
NM_001008546.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033329.mm.1 |
NM_001008546.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033329.mm.1 |
NM_145556.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033329.mm.1 |
NM_145556.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033329.mm.1 |
NR_027864.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033329.mm.1 |
NR_027864.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400033329.mm.1 |
TC0400004026.mm.1 |
MTA TC |