Assay Details
| Gene Symbol: Fbxo38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134136.3 | NP_598897.2 | 5 - 6 | 757 | 67 | |
| GenBank mRNA | AK031096.1 | - | 5 - 6 | 707 | 67 | |
| AK031347.1 | - | 5 - 6 | 688 | 67 | ||
| AK089046.1 | - | 5 - 6 | 729 | 67 | ||
| AY267463.1 | - | 5 - 6 | 690 | 67 | ||
| BC056348.1 | - | 5 - 6 | 709 | 5721407 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo38
Genomic Map