Assay Details
| Gene Symbol: Fbxo38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134136.3 | NP_598897.2 | 6 - 7 | 897 | 63 | |
| GenBank mRNA | AK031096.1 | - | 6 - 7 | 847 | 63 | |
| AK031347.1 | - | 6 - 7 | 828 | 63 | ||
| AK034761.1 | - | 1 - 2 | 81 | 63 | ||
| AK089046.1 | - | 6 - 7 | 869 | 63 | ||
| AY267463.1 | - | 6 - 7 | 830 | 63 | ||
| BC056348.1 | - | 6 - 7 | 849 | 5721407 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK034761.1 transcripts.
Set Membership: |
|

Fbxo38
Genomic Map