Assay Details
| Gene Symbol: Apitd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027263.2 | NP_081539.1 | 4 - 5 | 350 | 83 | |
| GenBank mRNA | AK011725.1 | - | 4 - 5 | 299 | 83 | |
| AK012995.1 | - | 4 - 5 | 332 | 83 | ||
| AK013038.1 | - | 4 - 5 | 332 | 83 | ||
| BC138926.1 | - | 4 - 5 | 291 | 8861948 | 83 | |
| BC138927.1 | - | 4 - 5 | 291 | 8861949 | 83 | |
| DV656255.1 | - | 4 - 5 | 350 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23398857 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23398855 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23398854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23398856 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270519 | AK012995.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270519 | AK013038.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270519 | NM_027263.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270519 | AK011725.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270519 | BC138926.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270519 | AK012995.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270519 | AK013038.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270519 | NM_027263.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270519 | AK011725.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270519 | BC138926.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P206058 | BC048521.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P206058 | BC048521.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472059 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 770550 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164285 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278899 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 959881 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168195 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162883 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270519 | AK012995.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270519 | AK013038.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270519 | NM_027263.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270519 | AK011725.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270519 | BC138926.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270519 | AK012995.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270519 | AK013038.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270519 | NM_027263.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270519 | AK011725.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270519 | BC138926.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23398821 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162883 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01015168 | - | - |
| Whole Mouse Genome G4122A | A_52_P206058 | BC048521.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P206058 | BC048521.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270519 | AK012995.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270519 | AK013038.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270519 | NM_027263.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270519 | AK011725.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270519 | BC138926.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270519 | AK012995.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270519 | AK013038.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270519 | NM_027263.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270519 | AK011725.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P270519 | BC138926.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104536 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162883 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03829611 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03959976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23398857 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23398855 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23398854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23398856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23398802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03959975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04008575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03358823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03433514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03817027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03817024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03203852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03317902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03317258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03663397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03946836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23398844 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10518566 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518562 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518563 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518565 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518567 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453067_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10518561 | - | - |
| Mouse Genome 430 2.0 Array | 1453067_at | - | - |
| Mouse Genome 430 2.0 Array | 1453067_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69928_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69928_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400033381.mm.1 | NM_027263.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033381.mm.1 | NM_027263.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033381.mm.1 | TC0400004032.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400033387.mm.1 | NM_027263.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033387.mm.1 | NM_027263.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033387.mm.1 | TC0400004032.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400033388.mm.1 | NM_027263.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033388.mm.1 | NM_027263.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033388.mm.1 | TC0400004032.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400033391.mm.1 | NM_027263.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033391.mm.1 | NM_027263.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033391.mm.1 | TC0400004032.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400033383.mm.1 | NM_027263.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033383.mm.1 | NM_027263.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033383.mm.1 | TC0400004032.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400033386.mm.1 | NM_027263.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033386.mm.1 | NM_027263.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033386.mm.1 | TC0400004032.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400033385.mm.1 | NM_027263.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033385.mm.1 | NM_027263.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033385.mm.1 | TC0400004032.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453067_PM_at | - | - |
| Mouse Expression Set 430 | 1453067_at | - | - |
| Murine Genome U74v2 Set | 167777_PM_r_at | - | - |
| Murine Genome U74v2 Set | 167777_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453067_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453067_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2260458 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002260458 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231416 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231416 | - | - |
| mouse-6 v1.1 (Illumina) | scl23502.6.1_40-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2260458 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23502.6.1_40-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260458 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2983714 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2983714 | - | - |

Apitd1
Genomic Map