Assay Details
| Gene Symbol: Lrp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080926.1 | NP_001074395.1 | 17 - 18 | 2550 | 63 | |
| NR_033496.1 | 18 - 19 | 2928 | 63 | |||
| GenBank mRNA | AJ312058.1 | - | 17 - 18 | 2396 | 63 | |
| BC127061.1 | - | 18 - 19 | 2303 | 63 | ||
| D85463.1 | - | 18 - 19 | 2926 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrp8
Genomic Map