Assay Details
| Gene Symbol: Cers2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029789.1 | NP_084065.1 | 10 - 11 | 1107 | 81 | |
| GenBank mRNA | AK002626.1 | - | 10 - 11 | 1074 | 81 | |
| AK050161.1 | - | 10 - 11 | 1088 | 81 | ||
| AK146106.1 | - | 10 - 11 | 1088 | 81 | ||
| AK159325.1 | - | 10 - 11 | 1086 | 81 | ||
| AY029532.1 | - | 10 - 11 | 1107 | 81 | ||
| BC006847.1 | - | 10 - 11 | 1079 | 3598473 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22343444 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22343446 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22343445 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22343442 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22343443 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P517381 | BC006847.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P517381 | AK159325.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P517381 | AY029532.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P517381 | AK050161.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P517381 | AK002626.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P517381 | BC006847.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P517381 | AK159325.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P517381 | AY029532.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P517381 | AK050161.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P517381 | AK002626.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 705645 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 317818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 132242 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908505 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53404 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179632 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P517381 | BC006847.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P517381 | AK159325.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P517381 | AY029532.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P517381 | AK050161.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P517381 | AK002626.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P517381 | BC006847.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P517381 | AK159325.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P517381 | AY029532.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P517381 | AK050161.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P517381 | AK002626.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517381 | BC006847.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517381 | AK159325.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517381 | AY029532.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517381 | AK050161.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517381 | AK002626.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517381 | BC006847.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517381 | AK159325.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517381 | AY029532.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517381 | AK050161.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517381 | AK002626.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22343451 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179632 | - | - |
| Whole Mouse Genome G4122A | A_51_P517381 | BC006847.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517381 | AK159325.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517381 | AY029532.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517381 | AK050161.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517381 | AK002626.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517381 | BC006847.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517381 | AK159325.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517381 | AY029532.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517381 | AK050161.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517381 | AK002626.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128066 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179632 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22343443 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22343445 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22343444 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22343446 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22343442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22343487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02838819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03025224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03179111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02972559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03050131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03025263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02674886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343428 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03025255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02764429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03179107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02375460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03025228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22343493 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10494234 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494236 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494228 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494230 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494235 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494231 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494229 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494237 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494233 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494232 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423955_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10494227 | - | - |
| Mouse Genome 430 2.0 Array | 1423955_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423955_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1420974_at | - | - |
| Mouse Genome 430 2.0 Array | 1420974_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76893_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76893_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300007095.mm.1 | NM_029789.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300007095.mm.1 | TC0300000897.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423955_a_at | - | - |
| Mouse Expression Set 430 | 1423955_PM_a_at | - | - |
| Murine Genome U74v2 Set | 160396_PM_at | - | - |
| Murine Genome U74v2 Set | 160396_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423955_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423955_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423955_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423955_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6770358 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4120086 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4120086 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6770358 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004120086 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006770358 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2653151 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229562 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2653151 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229562 | - | - |
| mouse-6 v1.1 (Illumina) | scl076893.11_90-S | - | - |
| mouse-6 v1.1 (Illumina) | scl22884.7.1_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6770358 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4120086 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22884.7.1_23-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl076893.11_90-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4120086 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6770358 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2893993 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2893992 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2893999 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2893999 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2893993 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2893992 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl076893.11_90-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl22884.7.1_23-S | - | - |

Cers2
Genomic Map